Anti-EPN2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: EPN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77854-836
- 77736-033
- Antigen Symbol:
- EPN2
- EPN2
- Antigen Synonyms:
- EHB21|EPN 2|EPN2_HUMAN|EPS 15 interacting protein 2|EPS-15-interacting protein 2|Eps15 binding protein|Epsin 2|Epsin-2|Epsin2|KIAA1065|OTTHUMP00000065808|OTTHUMP00000065809|OTTHUMP00000065810|OTTHUMP00000065811|OTTHUMP00000065889
- EHB21|EPN 2|EPN2_HUMAN|EPS 15 interacting protein 2|EPS-15-interacting protein 2|Eps15 binding protein|Epsin 2|Epsin-2|Epsin2|KIAA1065|OTTHUMP00000065808|OTTHUMP00000065809|OTTHUMP00000065810|OTTHUMP00000065811|OTTHUMP00000065889
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95208
- O95208
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Epsin-2 protein (15-300AA)
- Recombinant human Epsin-2 protein (15-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together