Anti-OLR1 Rabbit Polyclonal Antibody
77745-456
New Product
- Antigen Symbol:OLR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P78380
- Antigen Synonyms:C-type lectin domain family 8 member A| CLEC8A| hLOX 1| hLOX-1| Lectin like oxidized LDL receptor 1| Lectin like oxLDL receptor 1| Lectin type oxidized LDL receptor 1| Lectin-like oxidized LDL receptor 1| Lectin-like oxLDL receptor 1| Lectin-type oxidized LDL receptor 1| low density lipoprotein oxidized, receptor 1| LOX-1| LOXIN| OLR1_HUMAN| Ox LDL receptor 1| Ox-LDL receptor 1| Oxidised low density lipoprotein (lectin like) receptor 1| Oxidized low density lipoprotein receptor 1| Oxidized low density lipoprotein receptor 1 soluble form| Oxidized low-density lipoprotein receptor 1| OxLDL receptor 1 | SCARE1| Scavenger receptor class E, member 1| SLOX1| soluble form| SR-EI
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:SR-EI
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: OLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human