Anti-OPN3 Rabbit Polyclonal Antibody
Catalog # 77736-833
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OPN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q9H1Y3
- Antigen Synonyms:ECPN|Encephalopsin|ERO|NMO 1|NMO1|OPN 3|Opn3|OPN3_HUMAN|Opsin 3|Opsin-3|Opsin3|Panopsin
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:ECPN
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, IHC:1:100 to 1:300, IF:1:200 to 1:1000, ELISA:1:5000
Type:
Antigen: OPN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse