Anti-FAAH Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: FAAH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77855-079
- 77736-388
- Antigen Symbol:
- FAAH
- FAAH
- Antigen Synonyms:
- Anandamide amidohydrolase 1|Anandamide amidohydrolase|FA2H|FAAH|FAAH-1|FAAH1_HUMAN|Fatty acid 2-hydroxylase|Fatty acid alpha-hydroxylase|Fatty acid amide hydrolase|Fatty-acid amide hydrolase 1|MGC102823|MGC138146|Oleamide hydrolase 1|Oleamide hydrolase
- Anandamide amidohydrolase 1|Anandamide amidohydrolase|FA2H|FAAH|FAAH-1|FAAH1_HUMAN|Fatty acid 2-hydroxylase|Fatty acid alpha-hydroxylase|Fatty acid amide hydrolase|Fatty-acid amide hydrolase 1|MGC102823|MGC138146|Oleamide hydrolase 1|Oleamide hydrolase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00519
- O00519
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fatty-acid amide hydrolase 1 protein (205-342AA)
- Recombinant Human Fatty-acid amide hydrolase 1 protein (205-342AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together