Anti-ENOX1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ENOX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77854-747
- 77735-920
- Antigen Symbol:
- ENOX1
- ENOX1
- Antigen Synonyms:
- PIG38Ecto-NOX disulfide-thiol exchanger 1|Candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase|cCNOX|Cell proliferation-inducing gene 38 protein|Constitutive Ecto-NOX|cNOX) [Includes: Hydroquinone [NADH] oxidase|EC 1.-.-.-)|Protein disulfide-thiol oxidoreductase|EC 1.-.-.-)]
- PIG38Ecto-NOX disulfide-thiol exchanger 1|Candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase|cCNOX|Cell proliferation-inducing gene 38 protein|Constitutive Ecto-NOX|cNOX) [Includes: Hydroquinone [NADH] oxidase|EC 1.-.-.-)|Protein disulfide-thiol oxidoreductase|EC 1.-.-.-)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TC92
- Q8TC92
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ecto-NOX disulfide-thiol exchanger 1 protein (101-340AA)
- Recombinant Human Ecto-NOX disulfide-thiol exchanger 1 protein (101-340AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together