Anti-EIF4E3 Rabbit Polyclonal Antibody
Catalog # 77737-955
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EIF4E3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8N5X7
- Antigen Synonyms:EIF4E3Eukaryotic translation initiation factor 4E type 3|eIF-4E type 3|eIF-4E3|eIF4E type 3|eIF4E-3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:EIF4E3Eukaryotic translation initiation factor 4E type 3
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: EIF4E3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat