Anti-GABPB2 Rabbit Polyclonal Antibody
77737-820
New Product
- Antigen Symbol:GABPB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8TAK5
- Antigen Synonyms:BABPB2|E4TF1|E4TF1 47|E4TF1 53|E4TF1B|GA binding protein beta chain|GA binding protein transcription factor beta subunit 2|GA-binding protein subunit beta-2|GABP 2|GABP subunit beta 1|GABP subunit beta 2|GABP subunit beta-2|GABP2_HUMAN|GABPB 1|GABPB-2|GABPB1|NRF2B1|NRF2B2|Nuclear respiratory factor 2|Nuclear respiratory factor 2 subunit beta|RP11-68I18.1|Transcription factor E4TF1 47|Transcription factor E4TF1 53
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Transcription factor E4TF1 53
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: GABPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat