Anti-NASP Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: NASP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-936
- 77743-937
- Antigen Symbol:
- NASP
- NASP
- Antigen Synonyms:
- 5033430J04Rik|AI131596|AI317140|D4Ertd767e|DKFZp547F162|Epcs32|FLB7527|FLJ31599|FLJ35510|MGC19722|MGC20372|MGC2297|MGC93869|NASP|NASP_HUMAN|nuclear autoantigenic sperm protein (histone binding)|Nuclear autoantigenic sperm protein|PRO1999|RP23 233B9.9
- 5033430J04Rik|AI131596|AI317140|D4Ertd767e|DKFZp547F162|Epcs32|FLB7527|FLJ31599|FLJ35510|MGC19722|MGC20372|MGC2297|MGC93869|NASP|NASP_HUMAN|nuclear autoantigenic sperm protein (histone binding)|Nuclear autoantigenic sperm protein|PRO1999|RP23 233B9.9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49321
- P49321
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Nuclear autoantigenic sperm protein (519-788AA)
- Nuclear autoantigenic sperm protein (519-788AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together