Anti-FOXN1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-569
- 77737-449
- Antigen Symbol:
- FOXN1
- FOXN1
- Antigen Synonyms:
- FKHL20|Forkhead box N1|Forkhead box protein N1|FOXN 1|FOXN1_HUMAN|RONU|Rowett nude|Transcription factor winged-helix nude|WHN|Winged helix nude|Winged-helix transcription factor nude
- FKHL20|Forkhead box N1|Forkhead box protein N1|FOXN 1|FOXN1_HUMAN|RONU|Rowett nude|Transcription factor winged-helix nude|WHN|Winged helix nude|Winged-helix transcription factor nude
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15353
- O15353
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Forkhead box protein N1 protein (1-160AA)
- Recombinant Human Forkhead box protein N1 protein (1-160AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together