Anti-MYLK3 Rabbit Polyclonal Antibody
77743-765
New Product
- Antigen Symbol:MYLK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q32MK0
- Antigen Synonyms:caMLCK|cardiac MLCK|Cardiac MyBP C associated Ca/CaM kinase|Cardiac-MyBP-C-associated Ca/CaM kinase|MLC kinase|MLCK|MLCK2|Mylk3|MYLK3_HUMAN|Myosin light chain kinase 3|Putative myosin light chain kinase 3
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Putative myosin light chain kinase 3
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MYLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human