Anti-GRK2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-280
- 77738-708
- Antigen Symbol:
- GRK2
- Antigen Synonyms:
- ADRBK1|Adrenergic beta receptor kinase 1|ARBK1_HUMAN|BARK|BARK1|Beta adrenergic receptor kinase 1|Beta ARK 1|Beta ARK1|Beta-adrenergic receptor kinase 1|Beta-ARK-1|FLJ16718|G protein coupled receptor kinase 2|G-protein coupled receptor kinase 2|GRK2
- ADRBK1|Adrenergic beta receptor kinase 1|ARBK1_HUMAN|BARK|BARK1|Beta adrenergic receptor kinase 1|Beta ARK 1|Beta ARK1|Beta-adrenergic receptor kinase 1|Beta-ARK-1|FLJ16718|G protein coupled receptor kinase 2|G-protein coupled receptor kinase 2|GRK2
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P25098
- P25098
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Beta-adrenergic receptor kinase 1 protein (2-221AA)
- Recombinant Human Beta-adrenergic receptor kinase 1 protein (2-221AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together