Anti-MST1R Rabbit Polyclonal Antibody
Catalog # 77743-545
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MST1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q04912
- Antigen Synonyms:c met related tyrosine kinase| CD136| CD136 antigen| CDw136| Macrophage stimulating 1 receptor (c met related tyrosine kinase)| Macrophage stimulating 1 receptor| Macrophage stimulating protein receptor alpha chain| MACROPHAGE STIMULATING PROTEIN RECEPTOR| Macrophage stimulating protein receptor beta chain| Macrophage-Stimulating 1 Receptor (MST1R)| Macrophage-stimulating protein receptor beta chain| MSP receptor| MST1R variant RON30| MST1R variant RON62| NPCA3| p185 RON| p185-Ron| Protein-tyrosine kinase 8| PTK 8| ptk8| PTK8 protein tyrosine kinase 8| Recepteur d鈥檕rigine nantais (RON)| RON| RON protein tyrosine kinase| RON variant E2E3| RON_HUMAN| Soluble RON variant 1| Soluble RON variant 2| Soluble RON variant 3| Soluble RON variant 4| Stem cell derived tyrosine kinase
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Stem cell derived tyrosine kinase
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MST1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat