Anti-HNRNPU Rabbit Polyclonal Antibody
Catalog # 77740-256
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HNRNPU
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q00839
- Antigen Synonyms:Heterogeneous nuclear ribonucleoprotein U|hnRNP U|hnRNP U protein|hnRNPU protein|HNRPU|HNRPU_HUMAN|p120|p120 nuclear protein|pp120|SAF A|SAF-A|SAFA|Scaffold attachment factor A|U21.1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:U21.1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HNRNPU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat