Anti-IFNGR2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFNGR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-710
- 77857-912
- Antigen Symbol:
- IFNGR2
- IFNGR2
- Antigen Synonyms:
- AF 1|AF-1|AF1|IFGR 2|IFGR2|IFN gamma Receptor beta|IFN-gamma receptor 2|IFN-gamma-R2|IFNGR 2|IFNGT 1|IFNGT1|IMD28|INGR2_HUMAN|Interferon gamma receptor 2 (interferon gamma transducer 1)|Interferon gamma receptor 2|Interferon gamma receptor accessory factor 1|Interferon gamma receptor beta chain|Interferon gamma receptor chain 2|Interferon gamma transducer 1
- AF 1|AF-1|AF1|IFGR 2|IFGR2|IFN gamma Receptor beta|IFN-gamma receptor 2|IFN-gamma-R2|IFNGR 2|IFNGT 1|IFNGT1|IMD28|INGR2_HUMAN|Interferon gamma receptor 2 (interferon gamma transducer 1)|Interferon gamma receptor 2|Interferon gamma receptor accessory factor 1|Interferon gamma receptor beta chain|Interferon gamma receptor chain 2|Interferon gamma transducer 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P38484
- P38484
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon gamma receptor 2 protein (28-205AA)
- Recombinant Human Interferon gamma receptor 2 protein (28-205AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together