Anti-NDUFS4 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFS4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-224
- 77744-225
- Antigen Symbol:
- NDUFS4
- NDUFS4
- Antigen Synonyms:
- AQDQ|CI 18|CI 18 kDa|CI AQDQ|CI-18 kDa|CI-AQDQ|Complex I 18 kDa|Complex I AQDQ|Complex I-18 kDa|Complex I-AQDQ|mitochondrial|mitochondrial respiratory chain complex I (18 KD subunit)|NADH coenzyme Q reductase|NADH dehydrogenase (ubiquinone) Fe S protein 4 18kDa|NADH dehydrogenase [ubiquinone] iron-sulfur protein 4|NADH dehydrogenase|NADH ubiquinone oxidoreductase 18 kDa subunit|NADH-ubiquinone oxidoreductase 18 kDa subunit|NDUS4_HUMAN
- AQDQ|CI 18|CI 18 kDa|CI AQDQ|CI-18 kDa|CI-AQDQ|Complex I 18 kDa|Complex I AQDQ|Complex I-18 kDa|Complex I-AQDQ|mitochondrial|mitochondrial respiratory chain complex I (18 KD subunit)|NADH coenzyme Q reductase|NADH dehydrogenase (ubiquinone) Fe S protein 4 18kDa|NADH dehydrogenase [ubiquinone] iron-sulfur protein 4|NADH dehydrogenase|NADH ubiquinone oxidoreductase 18 kDa subunit|NADH-ubiquinone oxidoreductase 18 kDa subunit|NDUS4_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43181
- O43181
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial protein (43-161AA)
- NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial protein (43-161AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together