Anti-GPN1 Rabbit Polyclonal Antibody
77738-765
New Product
- Antigen Symbol:GPN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9HCN4
- Antigen Synonyms:ATP(GTP) binding protein|ATPBD1A|FLJ51176|GPN loop GTPase 1|GPN-loop GTPase 1|GPN1_HUMAN|MBD2 interacting protein|MBD2-interacting protein|MBDin|NTPBP|RNA polymerase II associated protein 4|RPAP4|XAB1|XPA binding protein 1|XPA binding protein 1 GTPase|XPA-binding protein 1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:XPA-binding protein 1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: GPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat