Anti-FTSJ1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FTSJ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-638
- 77737-695
- Antigen Symbol:
- FTSJ1
- FTSJ1
- Antigen Synonyms:
- CDLIV|FTSJ 1|FtsJ homolog 1 (E. coli)|FtsJ homolog 1|FtsJ RNA methyltransferase homolog 1|JM23|Mental retardation X linked 44|Mental retardation X linked 9|MRX44|MRX9|Protein ftsJ homolog 1|Putative ribosomal RNA methyltransferase 1|RRMJ1|RRMJ1_HUMAN|rRNA (uridine 2' O ) methyltransferase|rRNA (uridine-2''-O-)-methyltransferase|SPB1|TRM7
- CDLIV|FTSJ 1|FtsJ homolog 1 (E. coli)|FtsJ homolog 1|FtsJ RNA methyltransferase homolog 1|JM23|Mental retardation X linked 44|Mental retardation X linked 9|MRX44|MRX9|Protein ftsJ homolog 1|Putative ribosomal RNA methyltransferase 1|RRMJ1|RRMJ1_HUMAN|rRNA (uridine 2' O ) methyltransferase|rRNA (uridine-2''-O-)-methyltransferase|SPB1|TRM7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UET6
- Q9UET6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Putative tRNA (cytidine(32)/guanosine(34)-2\\\'-O)-methyltransferase protein (1-329AA)
- Recombinant Human Putative tRNA (cytidine(32)/guanosine(34)-2\\\'-O)-methyltransferase protein (1-329AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together