Anti-EXOC2 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: EXOC2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-276
- 77854-997
- Antigen Symbol:
- EXOC2
- EXOC2
- Antigen Synonyms:
- EXOC2_HUMAN| Exocyst complex component 2| Exocyst complex component Sec5| FLJ11026 | SEC5| SEC5 like 1| SEC5L1| Sec5p
- EXOC2_HUMAN|Exocyst complex component 2|Exocyst complex component Sec5|FLJ11026 |SEC5|SEC5 like 1|SEC5L1|Sec5p
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96KP1
- Q96KP1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Exocyst complex component 2 protein (1-220AA)
- Recombinant Human Exocyst complex component 2 protein (1-220AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together