Anti-MAP3K2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP3K2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-393
- 77742-667
- Antigen Symbol:
- MAP3K2
- MAP3K2
- Antigen Synonyms:
- M3K2_HUMAN|MAPK/ERK kinase kinase 2|MEK kinase 2|MEKK 2|MEKK2b|Mitogen activated protein kinase kinase kinase 2|Mitogen-activated protein kinase kinase kinase 2
- M3K2_HUMAN|MAPK/ERK kinase kinase 2|MEK kinase 2|MEKK 2|MEKK2b|Mitogen activated protein kinase kinase kinase 2|Mitogen-activated protein kinase kinase kinase 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2U5
- Q9Y2U5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitogen-activated protein kinase kinase kinase 2 protein (357-619AA)
- Recombinant Human Mitogen-activated protein kinase kinase kinase 2 protein (357-619AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together