Anti-RPA2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RPA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-097
- 77744-096
- Antigen Symbol:
- RPA2
- RPA2
- Antigen Synonyms:
- 60S acidic ribosomal protein P1|AA409079|AI325195|AU020965|ik:tdsubc_2g1|M(2)21C|MGC137236|OTTHUMP00000004008|p32|p34|RCJMB04_6d17 replication protein A2|32kDa|REPA2|Replication factor A protein 2|Replication protein A 32 kDa subunit|Replication protein A 32kDa subunit|Replication protein A 34 kDa subunit|Replication protein A|Replication Protein A2 (32kDa)|Replication protein A2|RF-A protein 2|Rf-A2|RFA|RFA2_HUMAN|RP-A p32|RP-A p34|RP21C|RPA 2|RPA 32|RPA|Rpa2|RPA32|RPA34|RpLP1|RpP2|xx:tdsubc_2g1|zgc:109822
- 60S acidic ribosomal protein P1|AA409079|AI325195|AU020965|ik:tdsubc_2g1|M(2)21C|MGC137236|OTTHUMP00000004008|p32|p34|RCJMB04_6d17 replication protein A2|32kDa|REPA2|Replication factor A protein 2|Replication protein A 32 kDa subunit|Replication protein A 32kDa subunit|Replication protein A 34 kDa subunit|Replication protein A|Replication Protein A2 (32kDa)|Replication protein A2|RF-A protein 2|Rf-A2|RFA|RFA2_HUMAN|RP-A p32|RP-A p34|RP21C|RPA 2|RPA 32|RPA|Rpa2|RPA32|RPA34|RpLP1|RpP2|xx:tdsubc_2g1|zgc:109822
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15927
- P15927
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Replication protein A 32 kDa subunit protein (1-267AA)
- Replication protein A 32 kDa subunit protein (1-267AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together