Anti-HIST1H1D Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:1 to 1:10
Type:
Antigen: HIST1H1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-747
- 77856-942
- Antigen Symbol:
- HIST1H1D
- Antigen Synonyms:
- H1 histone family member 3|H1.3|H13_HUMAN|H1F3|HIST1 H1D|HIST1H1D|Histone 1 H1d|Histone cluster 1 H1d|Histone H1.3|Histone H1c|MGC138176
- H1 histone family member 3|H1.3|H13_HUMAN|H1F3|HIST1 H1D|HIST1H1D|Histone 1 H1d|Histone cluster 1 H1d|Histone H1.3|Histone H1c|MGC138176
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P16402
- P16402
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoFluorescence:
- Yes
- ChIP:
- Yes
- Yes
- Immunogen:
- Peptide sequence around site of Thr (179) derived from Human Histone H1.3
- Peptide sequence around site of Thr (179) derived from Human Histone H1.3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together