Anti-LEFTY2 Rabbit Polyclonal Antibody
Catalog # 77742-075
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:LEFTY2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O00292
- Antigen Synonyms:EBAF|Endometrial bleeding associated factor (left right determination factor A transforming growth factor beta superfamily)|Endometrial bleeding associated factor|Endometrial bleeding-associated factor|Left right determination factor 2|Left right determination factor A|Left-right determination factor 2|Left-right determination factor A|LEFTA|LEFTY 2|LEFTYA|LFTY2 transforming growth factor|beta -4|LFTY2|mouse|homolog of|LFTY2_HUMAN|MGC46222|Protein lefty 2|Protein lefty A|Protein lefty-2|Protein lefty-A|Protein PSEC0024|TGF beta 4|TGF-beta-4|TGFB4|Transforming growth factor beta 4|Transforming growth factor beta-4
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Transforming growth factor beta-4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: LEFTY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat