Anti-RERE Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: RERE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77857-971
- 77740-890
- Antigen Symbol:
- RERE
- RERE
- Antigen Synonyms:
- 1110033A15Rik|AI414665|ARG|arginine glutamic acid dipeptide (RE) repeats|arginine glutamic acid dipeptide repeats|Arginine glutamic acid dipeptide repeats protein|Arginine-glutamic acid dipeptide repeats protein|ARGININE-GLUTAMIC ACID REPEATS-ENCODING GENE|ARP|ATN1L|Atr2|atrophin 1 like|Atrophin 1 like protein|Atrophin 1 related protein|atrophin 2|atrophin related protein|ATROPHIN-1-ASSOCIATED PROTEIN|Atrophin-1-like protein|Atrophin-1-related protein|atrophin-2|ATROPHIN-RELATED PROTEIN|AW742570|DNB1|FLJ38775|KIAA0458|mKIAA0458|NEDBEH|OTTHUMP00000001701|RERE_HUMAN
- 1110033A15Rik|AI414665|ARG|arginine glutamic acid dipeptide (RE) repeats|arginine glutamic acid dipeptide repeats|Arginine glutamic acid dipeptide repeats protein|Arginine-glutamic acid dipeptide repeats protein|ARGININE-GLUTAMIC ACID REPEATS-ENCODING GENE|ARP|ATN1L|Atr2|atrophin 1 like|Atrophin 1 like protein|Atrophin 1 related protein|atrophin 2|atrophin related protein|ATROPHIN-1-ASSOCIATED PROTEIN|Atrophin-1-like protein|Atrophin-1-related protein|atrophin-2|ATROPHIN-RELATED PROTEIN|AW742570|DNB1|FLJ38775|KIAA0458|mKIAA0458|NEDBEH|OTTHUMP00000001701|RERE_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P2R6
- Q9P2R6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human RERE
- Synthetic peptide of Human RERE
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together