Anti-ACTB Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACTB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-373
- 77745-372
- Antigen Symbol:
- ACTB
- ACTB
- Antigen Synonyms:
- A26C1A|A26C1B|ACTB|ACTB_HUMAN|Actin beta|Actin cytoplasmic 1|Actin|cytoplasmic 1|N-terminally processed|Actx|b actin|b-actin|Beta cytoskeletal actin|Beta-actin|BRWS1|E430023M04Rik|Melanoma X actin|MGC128179|PS1TP5 binding protein 1|PS1TP5BP1
- A26C1A|A26C1B|ACTB|ACTB_HUMAN|Actin beta|Actin cytoplasmic 1|Actin|cytoplasmic 1|N-terminally processed|Actx|b actin|b-actin|Beta cytoskeletal actin|Beta-actin|BRWS1|E430023M04Rik|Melanoma X actin|MGC128179|PS1TP5 binding protein 1|PS1TP5BP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P60709
- P60709
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Actin, cytoplasmic 1 protein (102-375AA)
- Actin, cytoplasmic 1 protein (102-375AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together