Anti-MAPK6 Rabbit Recombinant polyclonal Antibody
New Product
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: MAPK6
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-467
- 77742-751
- Antigen Symbol:
- MAPK6
- MAPK6
- Antigen Synonyms:
- ERK-3|ERK3|Extracellular signal regulated kinase 3|Extracellular signal regulated kinase p97|Extracellular signal-regulated kinase 3|HsT17250|MAP kinase 6|MAP kinase isoform p97|MAPK 6|Mitogen activated protein kinase 6|Mitogen-activated protein kinase 6|MK06_HUMAN|p97 MAPK|p97-MAPK|PRKM6|Protein kinase mitogen activated 5|Protein kinase mitogen activated 6
- ERK-3| ERK3| Extracellular signal regulated kinase 3| Extracellular signal regulated kinase p97| Extracellular signal-regulated kinase 3| HsT17250| MAP kinase 6| MAP kinase isoform p97| MAPK 6| Mitogen activated protein kinase 6| Mitogen-activated protein kinase 6| MK06_HUMAN| p97 MAPK| p97-MAPK| PRKM6| Protein kinase mitogen activated 5| Protein kinase mitogen activated 6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16659
- Q16659
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitogen-activated protein kinase 6 protein (383-712AA)
- Recombinant Human Mitogen-activated protein kinase 6 protein (383-712AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together