Anti-APOBEC4 Rabbit Polyclonal Antibody
Catalog # 77738-963
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:APOBEC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8WW27
- Antigen Synonyms:ABEC4_HUMAN|APOBEC 4|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4|Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 4|C1orf169|MGC26594|Putative C->U editing enzyme Putative C->U-editing enzyme APOBEC-4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:ABEC4_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: APOBEC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human