Anti-Nid1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Nid1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77744-444
- 77744-443
- Antigen Symbol:
- Nid1
- Nid1
- Antigen Synonyms:
- Nid1|Ent|Nidogen-1|NID-1|Entactin
- Nid1|Ent|Nidogen-1|NID-1|Entactin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10493
- P10493
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nidogen-1 protein (428-665AA)
- Nidogen-1 protein (428-665AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together