Anti-NOS2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NOS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-659
- 77744-660
- Antigen Symbol:
- NOS2
- NOS2
- Antigen Synonyms:
- HEP-NOS|Hepatocyte NOS|HEPNOS|inducible|Inducible nitric oxide synthase|Inducible NO synthase|Inducible NOS|iNOS|MAC NOS|Macrophage NOS|Nitric oxide synthase 2 inducible|Nitric oxide synthase 2 inducible macrophage|nitric oxide synthase 2A (inducible|hepatocytes)|Nitric oxide synthase|Nitric oxide synthase inducible|nitric oxide synthase|macrophage|NOS 2|NOS|Nos II|NOS type II|nos2|NOS2_HUMAN|NOS2A|Inducible|Hepatocyte|Peptidyl-cysteine S-nitrosylase NOS2
- HEP-NOS|Hepatocyte NOS|HEPNOS|inducible|Inducible nitric oxide synthase|Inducible NO synthase|Inducible NOS|iNOS|MAC NOS|Macrophage NOS|Nitric oxide synthase 2 inducible|Nitric oxide synthase 2 inducible macrophage|nitric oxide synthase 2A (inducible|hepatocytes)|Nitric oxide synthase|Nitric oxide synthase inducible|nitric oxide synthase|macrophage|NOS 2|NOS|Nos II|NOS type II|nos2|NOS2_HUMAN|NOS2A|Inducible|Hepatocyte|Peptidyl-cysteine S-nitrosylase NOS2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35228
- P35228
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nitric oxide synthase, inducible protein (17-141AA)
- Nitric oxide synthase, inducible protein (17-141AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together