Anti-KIAA0391 Rabbit Polyclonal Antibody
Catalog # 77741-577
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KIAA0391
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O15091
- Antigen Synonyms:KIAA0391 gene| Mitochondrial ribonuclease P protein 3| Mitochondrial RNase P protein 3| Mitochondrial RNase P subunit 3| MRPP3| MRRP3_HUMAN| PRORP| Proteinaceous RNase P
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Proteinaceous RNase P
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KIAA0391
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat