Anti-NAT1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: NAT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-941
- 77743-940
- Antigen Symbol:
- NAT1
- NAT1
- Antigen Synonyms:
- AAC1|ARY1_HUMAN|Arylamide acetylase 1|Arylamine N acetyltransferase 1|Arylamine N-acetyltransferase 1|mNAT|Monomorphic arylamine N acetyltransferase|Monomorphic arylamine N-acetyltransferase|N acetyltransferase type 1|N-acetyltransferase type 1|NAT 1|NAT-1|nat1|NATI
- AAC1|ARY1_HUMAN|Arylamide acetylase 1|Arylamine N acetyltransferase 1|Arylamine N-acetyltransferase 1|mNAT|Monomorphic arylamine N acetyltransferase|Monomorphic arylamine N-acetyltransferase|N acetyltransferase type 1|N-acetyltransferase type 1|NAT 1|NAT-1|nat1|NATI
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P18440
- P18440
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Arylamine N-acetyltransferase 1 protein (1-288AA)
- Arylamine N-acetyltransferase 1 protein (1-288AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together