Order Entry
United States
ContactUsLinkComponent
Anti-MEIS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MEIS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MEIS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77859-653
  • 77742-994
  • Antigen Symbol:
  • MEIS2
  • MEIS2
  • Antigen Synonyms:
  • Homeobox protein HsT18361|Meis (mouse) homolog 2|Meis homeobox 2|Meis homolog 2|Meis1 myeloid ecotropic viral integration site 1 homolog 2 (mouse)|Meis1 myeloid ecotropic viral integration site 1 homolog 2|Meis1 related gene 1|Meis1 related protein 1|Meis1-related protein 1|MEIS2_HUMAN|MGC2820|MRG1|TALE homeobox protein Meis2
  • Homeobox protein HsT18361|Meis (mouse) homolog 2|Meis homeobox 2|Meis homolog 2|Meis1 myeloid ecotropic viral integration site 1 homolog 2 (mouse)|Meis1 myeloid ecotropic viral integration site 1 homolog 2|Meis1 related gene 1|Meis1 related protein 1|Meis1-related protein 1|MEIS2_HUMAN|MGC2820|MRG1|TALE homeobox protein Meis2
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O14770
  • O14770
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Homeobox protein Meis2 protein (377-470AA)
  • Recombinant Human Homeobox protein Meis2 protein (377-470AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together