Anti-NFE2L1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NFE2L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-380
- 77744-379
- Antigen Symbol:
- NFE2L1
- NFE2L1
- Antigen Synonyms:
- erythroid derived 2|like 1|Locus control region factor 1|Locus control region-factor 1|NF-E2-related factor 1|NF2L1_HUMAN|NFE2 related factor 1|NFE2-related factor 1|NRF1|Nuclear factor|Nuclear Factor Erythroid 2 Related Factor 1|Nuclear factor erythroid 2-related factor 1|Nuclear factor erythroid derived 2 like 1|TCF-11|TCF11|Transcription factor 11|Transcription factor HBZ17|Transcription factor LCR F1|Transcription factor LCR-F1
- erythroid derived 2|like 1|Locus control region factor 1|Locus control region-factor 1|NF-E2-related factor 1|NF2L1_HUMAN|NFE2 related factor 1|NFE2-related factor 1|NRF1|Nuclear factor|Nuclear Factor Erythroid 2 Related Factor 1|Nuclear factor erythroid 2-related factor 1|Nuclear factor erythroid derived 2 like 1|TCF-11|TCF11|Transcription factor 11|Transcription factor HBZ17|Transcription factor LCR F1|Transcription factor LCR-F1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14494
- Q14494
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Endoplasmic reticulum membrane sensor NFE2L1 protein (251-550AA)
- Endoplasmic reticulum membrane sensor NFE2L1 protein (251-550AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together