Anti-TAB1 Rabbit Polyclonal Antibody
Catalog # 77742-688
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q15750
- Antigen Synonyms:2310012M03Rik|3'-Tab1|MAP3K7IP 1|MAP3K7IP1|MGC57664|Mitogen activated protein kinase kinase kinase 7 interacting protein 1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 1|TAB 1|TAB1|TAB1_HUMAN|TAK1 binding protein 1|TAK1-binding protein 1|TGF beta activated kinase 1 binding protein 1|TGF-beta activated kinase 1/MAP3K7 binding protein 1|TGF-beta-activated kinase 1 and MAP3K7-binding protein 1|TGF-beta-activated kinase 1-binding protein 1|Transforming growth factor beta activated kinase binding protein 1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MAP3K7IP1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat