Anti-IL1R2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000
Type:
Antigen: IL1R2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77858-035
- 77740-979
- Antigen Symbol:
- IL1R2
- Antigen Synonyms:
- IL1 Receptor II|Antigen CDw121b|CD121|CD121 antigen-like family member B|CD121b|CD121B Antigen|CDw121b|IL 1 type II receptor|IL-1R-2|IL-1R-beta|IL-1R2|IL-1RT-2|IL-1RT2|IL1R Beta|Il1r-2|IL1R2|IL1R2_HUMAN|IL1R2c|IL1RB|Interleukin 1 receptor type II variant 3|Interleukin 1 receptor|beta|type II|Interleukin-1 receptor beta|Interleukin-1 receptor type 2|Interleukin-1 receptor type II|MGC137383|MGC47725|OTTHUMP00000207969|p68|Type II interleukin-1 receptor
- IL1 Receptor II|Antigen CDw121b|CD121|CD121 antigen-like family member B|CD121b|CD121B Antigen|CDw121b|IL 1 type II receptor|IL-1R-2|IL-1R-beta|IL-1R2|IL-1RT-2|IL-1RT2|IL1R Beta|Il1r-2|IL1R2|IL1R2_HUMAN|IL1R2c|IL1RB|Interleukin 1 receptor type II variant 3|Interleukin 1 receptor|beta|type II|Interleukin-1 receptor beta|Interleukin-1 receptor type 2|Interleukin-1 receptor type II|MGC137383|MGC47725|OTTHUMP00000207969|p68|Type II interleukin-1 receptor
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- P27930
- P27930
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Interleukin-1 receptor type 2 protein (20-320AA)
- Recombinant Human Interleukin-1 receptor type 2 protein (20-320AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together