Anti-GTF2IRD1 Rabbit Polyclonal Antibody
Catalog # 77739-243
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GTF2IRD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UHL9
- Antigen Synonyms:CREAM1|General transcription factor II I repeat domain containing protein 1|General transcription factor II-I repeat domain-containing protein 1|General transcription factor III|GT2D1_HUMAN|GTF2I repeat domain containing protein 1|GTF2I repeat domain-containing protein 1|GTF3|Muscle TFII I repeat domain-containing protein 1|Muscle TFII-I repeat domain-containing protein 1|MUSTRD1|MusTRD1/BEN|RBAP2|Slow muscle fiber enhancer binding protein|Slow-muscle-fiber enhancer-binding protein|USE B1 binding protein|USE B1-binding protein|WBSCR11|WBSCR12|Williams Beuren syndrome chromosome region 11 protein|Williams-Beuren syndrome chromosomal region 11 protein|Williams-Beuren syndrome chromosomal region 12 protein
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Williams-Beuren syndrome chromosomal region 12 protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GTF2IRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat