Anti-FDPS Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
0
Type:
Antigen: FDPS
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77737-019
- 77855-282
- Antigen Symbol:
- FDPS
- FDPS
- Antigen Synonyms:
- (2E| (2E,6E) farnesyl diphosphate synthase| 6E)-farnesyl diphosphate synthase| Dimethylallyltranstransferase| Farnesyl diphosphate synthase| Farnesyl diphosphate synthetase| Farnesyl pyrophosphate synthase| Farnesyl pyrophosphate synthetase| FPP synthase| FPP synthetase| FPPS| FPPS_HUMAN| FPS| Geranyltranstransferase
- (2E|(2E,6E) farnesyl diphosphate synthase|6E)-farnesyl diphosphate synthase|Dimethylallyltranstransferase|Farnesyl diphosphate synthase|Farnesyl diphosphate synthetase|Farnesyl pyrophosphate synthase|Farnesyl pyrophosphate synthetase|FPP synthase|FPP synthetase|FPPS|FPPS_HUMAN|FPS|Geranyltranstransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14324
- P14324
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Farnesyl pyrophosphate synthase protein (1-419AA)
- Recombinant Human Farnesyl pyrophosphate synthase protein (1-419AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together