Anti-LRBA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LRBA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-125
- 77742-340
- Antigen Symbol:
- LRBA
- LRBA
- Antigen Synonyms:
- Beige-like protein|BGL|C80285|CDC4-like protein|CDC4L|CVID8|D3Ertd775e|Lab300|LBA|Lipopolysaccharide-responsive and beige-like anchor protein|LPS-responsive vesicle trafficking beach and anchor containing|LRBA_HUMAN|Vesicle trafficking beach and anchor containing
- Vesicle trafficking beach and anchor containing|LRBA_HUMAN|BGL|LPS-responsive vesicle trafficking beach and anchor containing|CDC4L|LBA|CVID8|Lab300|Beige-like protein|C80285|CDC4-like protein|D3Ertd775e|Lipopolysaccharide-responsive and beige-like anchor protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50851
- P50851
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lipopolysaccharide-responsive and beige-like anchor protein (2-252AA)
- Recombinant Human Lipopolysaccharide-responsive and beige-like anchor protein (2-252AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together