Anti-IL1RAPL1 Rabbit Polyclonal Antibody
77740-980
New Product
- Antigen Symbol:IL1RAPL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NZN1
- Antigen Synonyms:IL 1 RAPL 1|IL 1RAPL 1|IL-1-RAPL-1|IL-1RAPL-1|IL1R8|IL1RAPL 1|IL1RAPL|IL1RAPL-1|IL1RAPL1|Interleukin 1 receptor 8|Interleukin 1 receptor accessory protein like 1|Interleukin-1 receptor accessory protein-like 1|IRPL1_HUMAN|Mental retardation X linked 10|Mental retardation X linked 21|Mental retardation X linked 34|MRX 21|MRX 34|MRX10|MRX21|MRX34|Oligophrenin 4|Oligophrenin-4|Oligophrenin4|OPHN 4|OPHN4|Three immunoglobulin domain containing IL 1 receptor related 2|Three immunoglobulin domain-containing IL-1 receptor-related 2|TIGIRR 2|TIGIRR-2|TIGIRR2|X linked interleukin 1 receptor accessory protein like 1|X-linked interleukin-1 receptor accessory protein-like 1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:X-linked interleukin-1 receptor accessory protein-like 1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: IL1RAPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat