Anti-ATP4B Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: ATP4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77745-661
- 77745-662
- Antigen Symbol:
- ATP4B
- ATP4B
- Antigen Synonyms:
- Gastric H(+)/K(+) ATPase subunit beta|ATP4B|ATP4B_HUMAN|ATP6B|ATPase H+/K+ exchanging beta polypeptide|ATPase H+/K+ transporting beta polypeptide|Gastric H K ATPase catalytic subunit|Gastric H+/K+ ATPase beta subunit|Gastric hydrogen potassium ATPase|Gastric hydrogen potassium ATPase beta|Hydrogen/potassium exchanging ATPase 4B|OTTHUMP00000178856|Parietal cell antigen|Potassium transporting ATPase beta chain|Potassium transporting ATPase subunit beta|Potassium-transporting ATPase subunit beta|Proton pump|Proton pump beta chain
- Gastric H(+)/K(+) ATPase subunit beta|ATP4B|ATP4B_HUMAN|ATP6B|ATPase H+/K+ exchanging beta polypeptide|ATPase H+/K+ transporting beta polypeptide|Gastric H K ATPase catalytic subunit|Gastric H+/K+ ATPase beta subunit|Gastric hydrogen potassium ATPase|Gastric hydrogen potassium ATPase beta|Hydrogen/potassium exchanging ATPase 4B|OTTHUMP00000178856|Parietal cell antigen|Potassium transporting ATPase beta chain|Potassium transporting ATPase subunit beta|Potassium-transporting ATPase subunit beta|Proton pump|Proton pump beta chain
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51164
- P51164
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of ATP4B
- Fusion protein of ATP4B
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together