Anti-MECP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MECP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-946
- 77859-621
- Antigen Symbol:
- MECP2
- MECP2
- Antigen Synonyms:
- AUTSX 3|AUTSX3|DKFZp686A24160|Mbd 5|Mbd5|MECP 2|MeCP 2 protein|MeCP-2 protein|MECP2_HUMAN|Methyl CpG binding protein 2 (Rett syndrome)|Methyl CpG binding protein 2|Methyl-CpG-binding protein 2|MRX 16|MRX 79|MRX16|MRX79|MRXS 13|MRXS13|MRXSL|PPMX|RS|RTS|RTT|WBP 10|WBP10
- AUTSX 3|AUTSX3|DKFZp686A24160|Mbd 5|Mbd5|MECP 2|MeCP 2 protein|MeCP-2 protein|MECP2_HUMAN|Methyl CpG binding protein 2 (Rett syndrome)|Methyl CpG binding protein 2|Methyl-CpG-binding protein 2|MRX 16|MRX 79|MRX16|MRX79|MRXS 13|MRXS13|MRXSL|PPMX|RS|RTS|RTT|WBP 10|WBP10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51608
- P51608
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Methyl-CpG-binding protein 2 protein (1-280AA)
- Recombinant human Methyl-CpG-binding protein 2 protein (1-280AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together