Anti-ITGAV Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: ITGAV
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-297
- 77741-302
- Antigen Symbol:
- ITGAV
- ITGAV
- Antigen Synonyms:
- antigen identified by monoclonal L230|CD 51|CD51|DKFZp686A08142|Integrin alpha five|integrin alpha V beta 3|Integrin alpha-5|integrin alpha-V|Integrin alpha-V light chain|integrin alphaVbeta3|integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51)|ITAV_HUMAN|MSK 8|Msk8|Vitronectin receptor subunit alpha|VNRA|VTNR
- antigen identified by monoclonal L230| CD 51| CD51| DKFZp686A08142| Integrin alpha five| integrin alpha V beta 3| Integrin alpha-5| integrin alpha-V| Integrin alpha-V light chain| integrin alphaVbeta3| integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51)| ITAV_HUMAN| MSK 8| Msk8| Vitronectin receptor subunit alpha| VNRA| VTNR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06756
- P06756
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Integrin alpha-V protein (775-885AA)
- Recombinant Human Integrin alpha-V protein (775-885AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together