Anti-Gdf5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:10 to 1:100
Type:
Antigen: Gdf5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77855-895
- 77738-119
- Antigen Symbol:
- Gdf5
- Gdf5
- Antigen Synonyms:
- Gdf5| Bmp14| Bp| Gdf-5Growth/differentiation factor 5| GDF-5| Bone morphogenetic protein 14| BMP-14
- Gdf5|Bmp14|Bp|Gdf-5Growth/differentiation factor 5|GDF-5|Bone morphogenetic protein 14|BMP-14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43027
- P43027
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse Growth/differentiation factor 5 protein (376-495AA)
- Recombinant Mouse Growth/differentiation factor 5 protein (376-495AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together