Anti-GIPC1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GIPC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-004
- 77738-274
- Antigen Symbol:
- GIPC1
- GIPC1
- Antigen Synonyms:
- C19orf3|GIPC|RGS19IP1PDZ domain-containing protein GIPC1|GAIP C-terminus-interacting protein|RGS-GAIP-interacting protein|RGS19-interacting protein 1|Synectin|Tax interaction protein 2|TIP-2
- C19orf3|GIPC|RGS19IP1PDZ domain-containing protein GIPC1|GAIP C-terminus-interacting protein|RGS-GAIP-interacting protein|RGS19-interacting protein 1|Synectin|Tax interaction protein 2|TIP-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14908
- O14908
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human PDZ domain-containing protein GIPC1 protein (7-74AA)
- Recombinant Human PDZ domain-containing protein GIPC1 protein (7-74AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together