Anti-MOAP1 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: MOAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-866
- 77743-277
- Antigen Symbol:
- MOAP1
- MOAP1
- Antigen Synonyms:
- MAP 1|MAP-1|MAP1|MOAP1_HUMAN|Modulator of apoptosis 1|Modulator of Apoptosis|Paraneoplastic antigen like 4|Paraneoplastic antigen Ma4|paraneoplastic Ma antigen family member 4|PNMA4
- MAP 1|MAP-1|MAP1|MOAP1_HUMAN|Modulator of apoptosis 1|Modulator of Apoptosis|Paraneoplastic antigen like 4|Paraneoplastic antigen Ma4|paraneoplastic Ma antigen family member 4|PNMA4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96BY2
- Q96BY2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Modulator of apoptosis 1 protein (1-351AA)
- Recombinant human Modulator of apoptosis 1 protein (1-351AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together