Anti-KLHL41 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KLHL41
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-392
- 77741-431
- Antigen Symbol:
- KLHL41
- KLHL41
- Antigen Synonyms:
- cb52|fb24f05|fc15e07|Gm112|KBTBA_HUMAN|Kbtbd10|Kel like protein 23|Kel-like protein 23|Kelch related protein 1|Kelch repeat and BTB domain-containing protein 10|Kelch-related protein 1|KRP1|MGC143407|Sarcosin|sbcb52|wufb24f05|wufc15e07
- cb52|fb24f05|fc15e07|Gm112|KBTBA_HUMAN|Kbtbd10|Kel like protein 23|Kel-like protein 23|Kelch related protein 1|Kelch repeat and BTB domain-containing protein 10|Kelch-related protein 1|KRP1|MGC143407|Sarcosin|sbcb52|wufb24f05|wufc15e07
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60662
- O60662
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Kelch-like protein 41 protein (1-300AA)
- Recombinant Human Kelch-like protein 41 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together