Anti-MAP3K11 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: MAP3K11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-867
- 77743-281
- Antigen Symbol:
- MAP3K11
- Antigen Synonyms:
- 2610017K16Rik|EC 2.7.11.25|M3K11_HUMAN|Map3k11|MEKK11|MGC17114|Mitogen-activated protein kinase kinase kinase 11|Mixed lineage kinase 3|Mixed lineage protein kinase 3|Mlk-3|Mlk3|Protein tyrosine kinase PTK1|PTK1|RHOE|SH3 domain containing proline rich kinase|SPRK|Src-homology 3 domain-containing proline-rich kinase
- 2610017K16Rik|EC 2.7.11.25|M3K11_HUMAN|Map3k11|MEKK11|MGC17114|Mitogen-activated protein kinase kinase kinase 11|Mixed lineage kinase 3|Mixed lineage protein kinase 3|Mlk-3|Mlk3|Protein tyrosine kinase PTK1|PTK1|RHOE|SH3 domain containing proline rich kinase|SPRK|Src-homology 3 domain-containing proline-rich kinase
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q16584
- Q16584
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoFluorescence:
- Yes
- Immunogen:
- Recombinant Human Mitogen-activated protein kinase kinase kinase 11 protein (536-693AA)
- Recombinant Human Mitogen-activated protein kinase kinase kinase 11 protein (536-693AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together