Anti-KDM5C Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: KDM5C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77741-398
- 77858-375
- Antigen Symbol:
- KDM5C
- KDM5C
- Antigen Synonyms:
- Jumonji AT rich interactive domain 1C|MRXSJ|DXS1272E|Lysine-specific demethylase 5C|Histone demethylase JARID1C|SmcX protein|JARID1C|Smcx homolog X chromosome|MRX13|MRXSCJ|XE169|AT rich interactive domain 1C (RBP2 like)|Jumonji/ARID domain-containing protein 1C|KDM5C_HUMAN|Protein Xe169|Selected cDNA on X|JmjC domain containing protein SMCX|Jumonji|SMCX|Lysine (K) specific demethylase 5C|rbp2 like protein|Xe169 protein|MRXJ|Smcy homolog X linked|Protein SmcX
- DXS1272E|Histone demethylase JARID1C|JARID1C|JmjC domain containing protein SMCX|Jumonji AT rich interactive domain 1C|Jumonji|AT rich interactive domain 1C (RBP2 like)|Jumonji/ARID domain-containing protein 1C|KDM5C_HUMAN|Lysine (K) specific demethylase 5C|Lysine-specific demethylase 5C|MRX13|MRXJ|MRXSCJ|MRXSJ|Protein SmcX|Protein Xe169|rbp2 like protein|Selected cDNA on X|SMCX|Smcx homolog X chromosome|SmcX protein|Smcy homolog X linked|XE169|Xe169 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41229
- P41229
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysine-specific demethylase 5C protein (45-324AA)
- Recombinant Human Lysine-specific demethylase 5C protein (45-324AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together