Anti-IRF8 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: IRF8
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-207
- 77741-201
- Antigen Symbol:
- IRF8
- IRF8
- Antigen Synonyms:
- H ICSBP|H-ICSBP|HGNC:5358|HICSBP|ICSBP 1|ICSBP|ICSBP1|Interferon consensus sequence binding protein 1|Interferon consensus sequence binding protein|Interferon consensus sequence-binding protein|Interferon regulatory factor 8|IRF 8|IRF-8|IRF8_HUMAN|MYLS
- HGNC:5358|Interferon consensus sequence binding protein 1|Interferon regulatory factor 8|ICSBP 1|IRF-8|H ICSBP|Interferon consensus sequence binding protein|IRF 8|IRF8_HUMAN|MYLS|H-ICSBP|HICSBP|ICSBP|ICSBP1|Interferon consensus sequence-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02556
- Q02556
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon regulatory factor 8 protein (137-426AA)
- Recombinant Human Interferon regulatory factor 8 protein (137-426AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together