Anti-HMGCS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HMGCS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-185
- 77857-493
- Antigen Symbol:
- HMGCS2
- HMGCS2
- Antigen Synonyms:
- 3 hydroxy 3 methylglutaryl Coenzyme A synthase 2 (mitochondrial)|3 hydroxy 3 methylglutaryl Coenzyme A synthase 2|3 hydroxy 3 methylglutaryl coenzyme A synthase|3-hydroxy-3-methylglutaryl coenzyme A synthase|HMCS2_HUMAN|HMG CoA synthase|HMG-CoA synthase|HMGCS 2|Hydroxymethylglutaryl CoA synthase|Hydroxymethylglutaryl CoA synthase mitochondrial|Hydroxymethylglutaryl-CoA synthase|mitochondrial
- 3 hydroxy 3 methylglutaryl Coenzyme A synthase 2 (mitochondrial)|3 hydroxy 3 methylglutaryl Coenzyme A synthase 2|3 hydroxy 3 methylglutaryl coenzyme A synthase|3-hydroxy-3-methylglutaryl coenzyme A synthase|HMCS2_HUMAN|HMG CoA synthase|HMG-CoA synthase|HMGCS 2|Hydroxymethylglutaryl CoA synthase|Hydroxymethylglutaryl CoA synthase mitochondrial|Hydroxymethylglutaryl-CoA synthase|mitochondrial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54868
- P54868
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hydroxymethylglutaryl-CoA synthase, mitochondrial protein (426-508AA)
- Recombinant Human Hydroxymethylglutaryl-CoA synthase, mitochondrial protein (426-508AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together