Anti-EPS15 Rabbit Polyclonal Antibody
77736-552
New Product
- Antigen Symbol:EPS15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P42566
- Antigen Synonyms:EPS15|AF1PEpidermal growth factor receptor substrate 15|Protein Eps15|Protein AF-1p
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Protein AF-1p
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:10000
Type:
Antigen: EPS15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat